Tisk…

Anti-CD8A Rat Monoclonal Antibody [clone: 2.43] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Dodavatel: Abnova
ABNOMAB12214EA 10550 CZK
ABNOMAB12214
Anti-CD8A Rat Monoclonal Antibody [clone: 2.43] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Protilátky
Rat monoclonal antibody raised against Cd8a.

Type: Secondary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: PerCP/Cy5.5
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR