Tisk…

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Dodavatel: Abnova
ABNOMAB12221EA 9980 CZK
ABNOMAB12221
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Protilátky
Mouse monoclonal antibody raised against CD19.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR