Tisk…

Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abnova
ABNOMAB12222EA 10550 CZK
ABNOMAB12222
Anti-CD19 Rat Monoclonal Antibody [clone: 1D3] (FITC (Fluorescein Isothiocyanate))
Protilátky
Rat monoclonal antibody raised against Cd19.

Type: Secondary
Antigen: Cd19
Clonality: Monoclonal
Clone: 1D3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR