Tisk…

Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin))

Dodavatel: Abnova
ABNOMAB12224EA 9980 CZK
ABNOMAB12224
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PE (Phycoerythrin))
Protilátky
Mouse monoclonal antibody raised against CD28.

Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR