Tisk…

Anti-PTPRC Mouse Monoclonal Antibody [clone: HI30] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abnova
ABNOMAB12226EA 9980 CZK
ABNOMAB12226
Anti-PTPRC Mouse Monoclonal Antibody [clone: HI30] (FITC (Fluorescein Isothiocyanate))
Protilátky
Mouse monoclonal antibody raised against PTPRC.

Type: Secondary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR