Tisk…

Anti-PTPRC Rat Monoclonal Antibody [clone: RA3-6B2] (APC (Allophycocyanin))

Dodavatel: Abnova
ABNOMAB12228EA 10550 CZK
ABNOMAB12228
Anti-PTPRC Rat Monoclonal Antibody [clone: RA3-6B2] (APC (Allophycocyanin))
Protilátky
Rat monoclonal antibody raised against PTPRC.

Type: Secondary
Antigen: Abelson murine leukemia virus-induced pre-B tumor cells.
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR