Tisk…

Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abnova
ABNOMAB12232EA 9980 CZK
ABNOMAB12232
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2] (FITC (Fluorescein Isothiocyanate))
Protilátky
Mouse monoclonal antibody raised against ICAM1.

Type: Secondary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 15.2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR