Tisk…

Anti-ENG Mouse Monoclonal Antibody [clone: SN6] (PE (Phycoerythrin))

Dodavatel: Abnova
ABNOMAB12233EA 11380 CZK
ABNOMAB12233
Anti-ENG Mouse Monoclonal Antibody [clone: SN6] (PE (Phycoerythrin))
Protilátky
Mouse monoclonal antibody raised against ENG.

Type: Secondary
Antigen: ENG
Clonality: Monoclonal
Clone: SN6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR