Tisk…

Anti-FAM44A Mouse Monoclonal Antibody [clone: 2A1]

Dodavatel: Abnova
ABNOH00259282-M06EA 10890 CZK
ABNOH00259282-M06
Anti-FAM44A Mouse Monoclonal Antibody [clone: 2A1]
Protilátky
Mouse monoclonal antibody raised against a full-length recombinant FAM44A.

Type: Primary
Antigen: FAM44A
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR