Tisk…

Anti-HBB Mouse Monoclonal Antibody [clone: 7B12]

Dodavatel: Abnova
ABNOH00003043-M02AEA 10890 CZK
ABNOH00003043-M02A
Anti-HBB Mouse Monoclonal Antibody [clone: 7B12]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant HBB.

Type: Primary
Antigen: HBB
Clonality: Monoclonal
Clone: 7B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR