Tisk…

Anti-CD274 Mouse Monoclonal Antibody [clone: 2E11]

Dodavatel: Abnova
ABNOH00029126-M05EA 10890 CZK
ABNOH00029126-M05
Anti-CD274 Mouse Monoclonal Antibody [clone: 2E11]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD274.

Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR