Tisk…

Anti-CD276 Mouse Monoclonal Antibody [clone: 4E1]

Dodavatel: Abnova
ABNOH00080381-M27EA 10890 CZK
ABNOH00080381-M27
Anti-CD276 Mouse Monoclonal Antibody [clone: 4E1]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD276.

Type: Primary
Antigen: CD276
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR