Tisk…

Anti-CD80 Mouse Monoclonal Antibody [clone: 7D3]

Dodavatel: Abnova
ABNOH00000941-M05EA 11230 CZK
ABNOH00000941-M05
Anti-CD80 Mouse Monoclonal Antibody [clone: 7D3]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD80.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 7D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR