Tisk…

Anti-PDCD1 Mouse Monoclonal Antibody [clone: 6B10]

Dodavatel: Abnova
ABNOH00005133-M03EA 10890 CZK
ABNOH00005133-M03
Anti-PDCD1 Mouse Monoclonal Antibody [clone: 6B10]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant PDCD1.

Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone: 6B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR