Tisk…

Anti-CTLA4 Mouse Monoclonal Antibody [clone: 5G6]

Dodavatel: Abnova
ABNOH00001493-M37EA 11230 CZK
ABNOH00001493-M37
Anti-CTLA4 Mouse Monoclonal Antibody [clone: 5G6]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CTLA4.

Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone: 5G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR