Tisk…

Anti-PDCD1LG2 Mouse Monoclonal Antibody [clone: 1F12]

Dodavatel: Abnova
ABNOH00080380-M17EA 10890 CZK
ABNOH00080380-M17
Anti-PDCD1LG2 Mouse Monoclonal Antibody [clone: 1F12]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant PDCD1LG2.

Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 1F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR