Tisk…

Anti-KIAA1217 Mouse Monoclonal Antibody [clone: 3B10]

Dodavatel: Abnova
ABNOH00056243-M02EA 10890 CZK
ABNOH00056243-M02
Anti-KIAA1217 Mouse Monoclonal Antibody [clone: 3B10]
Protilátky
Mouse monoclonal antibody raised against a full-length recombinant KIAA1217.

Type: Primary
Antigen: KIAA1217
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR