Tisk…

Anti-LENG1 Mouse Monoclonal Antibody [clone: 2E7]

Dodavatel: Abnova
ABNOH00079165-M02EA 10890 CZK
ABNOH00079165-M02
Anti-LENG1 Mouse Monoclonal Antibody [clone: 2E7]
Protilátky
Mouse monoclonal antibody raised against a full-length recombinant LENG1.

Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR