Tisk…

Anti-LTA Mouse Monoclonal Antibody [clone: 3A1]

Dodavatel: Abnova
ABNOH00004049-M47EA 10890 CZK
ABNOH00004049-M47
Anti-LTA Mouse Monoclonal Antibody [clone: 3A1]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant LTA.

Type: Primary
Antigen: LTA
Clonality: Monoclonal
Clone: 3A1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR