Tisk…

Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 1E1]

Dodavatel: Abnova
ABNOH00000944-M02EA 10890 CZK
ABNOH00000944-M02
Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 1E1]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF8.

Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR