Tisk…
Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 4A6]
Dodavatel: Abnova
ABNOH00000944-M07EA 11230 CZK
ABNOH00000944-M07
Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 4A6]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF8.
Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 4A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human