Tisk…

Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 8F4]

Dodavatel: Abnova
ABNOMAB1180-M06EA 10890 CZK
ABNOMAB1180-M06
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 8F4]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10A.

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 8F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR