Tisk…

Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 4F6]

Dodavatel: Abnova
ABNOH00008795-M09EA 10890 CZK
ABNOH00008795-M09
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 4F6]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR