Tisk…

Anti-CD28 Mouse Monoclonal Antibody [clone: 4B5]

Dodavatel: Abnova
ABNOH00000940-M22EA 11230 CZK
ABNOH00000940-M22
Anti-CD28 Mouse Monoclonal Antibody [clone: 4B5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD28.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR