Tisk…
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1B7]
Dodavatel: Abnova
ABNOH00008795-M11EA 11230 CZK
ABNOH00008795-M11
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1B7]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human