Tisk…

Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1H2]

Dodavatel: Abnova
ABNOH00008795-M04EA 11230 CZK
ABNOH00008795-M04
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1H2]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR