Tisk…
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 4E9]
Dodavatel: Abnova
ABNOH00008795-M06EA 11230 CZK
ABNOH00008795-M06
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 4E9]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human