Tisk…
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1E11]
Dodavatel: Abnova
ABNOMAB1232-M04EA 11580 CZK
ABNOMAB1232-M04
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 1E11]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat