Tisk…

Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 2F9]

Dodavatel: Abnova
ABNOMAB1232-M05EA 10890 CZK
ABNOMAB1232-M05
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 2F9]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10B.

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR