Tisk…

Anti-CD86 Mouse Monoclonal Antibody [clone: 1F8]

Dodavatel: Abnova
ABNOH00000942-M07EA 10890 CZK
ABNOH00000942-M07
Anti-CD86 Mouse Monoclonal Antibody [clone: 1F8]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD86.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR