Tisk…

Anti-LTBR Mouse Monoclonal Antibody [clone: 8C7]

Dodavatel: Abnova
ABNOH00004055-M08EA 11230 CZK
ABNOH00004055-M08
Anti-LTBR Mouse Monoclonal Antibody [clone: 8C7]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant LTBR.

Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 8C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR