Tisk…

Anti-LTBR Mouse Monoclonal Antibody [clone: 7E7]

Dodavatel: Abnova
ABNOH00004055-M07EA 11230 CZK
ABNOH00004055-M07
Anti-LTBR Mouse Monoclonal Antibody [clone: 7E7]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant LTBR.

Type: Primary
Antigen: LTBR
Clonality: Monoclonal
Clone: 7E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR