Tisk…

Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 5F8]

Dodavatel: Abnova
ABNOH00003604-M01EA 10890 CZK
ABNOH00003604-M01
Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 5F8]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF9.

Type: Primary
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: 5F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR