Tisk…

Anti-EDA Mouse Monoclonal Antibody [clone: 1F6]

Dodavatel: Abnova
ABNOH00001896-M02EA 11230 CZK
ABNOH00001896-M02
Anti-EDA Mouse Monoclonal Antibody [clone: 1F6]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant EDA.

Type: Primary
Antigen: EDA
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR