Tisk…

Anti-ICOSLG Mouse Monoclonal Antibody [clone: 3H8]

Dodavatel: Abnova
ABNOH00023308-M05EA 10890 CZK
ABNOH00023308-M05
Anti-ICOSLG Mouse Monoclonal Antibody [clone: 3H8]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant ICOSLG.

Type: Primary
Antigen: ICOSLG
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR