Tisk…
Anti-CD28 Mouse Monoclonal Antibody [clone: 1E5]
Dodavatel: Abnova
ABNOH00000940-M03EA 11230 CZK
ABNOH00000940-M03
Anti-CD28 Mouse Monoclonal Antibody [clone: 1E5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD28.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human