Tisk…

Anti-CD28 Mouse Monoclonal Antibody [clone: 4G11]

Dodavatel: Abnova
ABNOH00000940-M18EA 10890 CZK
ABNOH00000940-M18
Anti-CD28 Mouse Monoclonal Antibody [clone: 4G11]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant CD28.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR