Tisk…

Anti-CDCA1 Mouse Polyclonal Antibody

Dodavatel: Abnova
ABNOH00083540-B01PEA 11230 CZK
ABNOH00083540-B01P
Anti-CDCA1 Mouse Polyclonal Antibody
Protilátky
Mouse polyclonal antibody raised against a full-length human CDCA1 protein.

Type: Primary
Antigen: CDCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR