Tisk…

Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 3E3]

Dodavatel: Abnova
ABNOH00023495-M24EA 10890 CZK
ABNOH00023495-M24
Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 3E3]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF13B.

Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR