Tisk…

Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 2E3]

Dodavatel: Abnova
ABNOH00009966-M07EA 11230 CZK
ABNOH00009966-M07
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 2E3]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF15.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR