Tisk…
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 1C2]
Dodavatel: Abnova
ABNOH00009966-M10EA 11230 CZK
ABNOH00009966-M10
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 1C2]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFSF15.
Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human