Tisk…

Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 5A3]

Dodavatel: Abnova
ABNOH00008792-M04EA 11230 CZK
ABNOH00008792-M04
Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 5A3]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.

Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 5A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR