Tisk…

Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 4C10]

Dodavatel: Abnova
ABNOH00008792-M06EA 10890 CZK
ABNOH00008792-M06
Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 4C10]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.

Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 4C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR