Tisk…
Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 5C12]
Dodavatel: Abnova
ABNOH00008792-M10EA 11230 CZK
ABNOH00008792-M10
Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 5C12]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.
Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 5C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human