Tisk…
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 5B5]
Dodavatel: Abnova
ABNOH00008764-M07EA 11230 CZK
ABNOH00008764-M07
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 5B5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF14.
Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human