Tisk…

Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 3H5]

Dodavatel: Abnova
ABNOH00008764-M22EA 10890 CZK
ABNOH00008764-M22
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 3H5]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF14.

Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR