Tisk…

Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 4D8]

Dodavatel: Abnova
ABNOH00027242-M48EA 10890 CZK
ABNOH00027242-M48
Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 4D8]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF21.

Type: Primary
Antigen: TNFRSF21
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR