Tisk…
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 3A1]
Dodavatel: Abnova
ABNOMAB1300-M01EA 11580 CZK
ABNOMAB1300-M01
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 3A1]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10C.
Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat