Tisk…

Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 4D12]

Dodavatel: Abnova
ABNOMAB1300-M03EA 11580 CZK
ABNOMAB1300-M03
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: 4D12]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10C.

Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR