Tisk…

Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 1E2]

Dodavatel: Abnova
ABNOMAB1297-M05EA 11580 CZK
ABNOMAB1297-M05
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 1E2]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10A.

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR